siRNA Details
Gene Symbol: IGSF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007237.2 | NP_001007238.1 | 10 | 3749 | ||
NM_001542.3 | NP_001533.2 | 10 | 3671 | |||
XM_005270793.2 | XP_005270850.1 | 11 | 3781 | |||
XM_005270794.4 | XP_005270851.1 | 9 | 3019 | |||
XM_006710593.3 | XP_006710656.1 | 10 | 3082 | |||
XM_011541315.1 | XP_011539617.1 | 11 | 3833 | |||
XM_011541316.2 | XP_011539618.1 | 5 | 2562 | |||
GenBank mRNA | AB007935.2 | - | 9 | 3081 | ||
AF031174.1 | - | 10 | 3146 | |||
BC110651.1 | - | 10 | 3749 | 6166346 | ||
BC130279.1 | - | 9 | 3419 | 40146310 | ||
BC144132.1 | - | 9 | 3419 | 9052652 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
