siRNA Details
Gene Symbol: LINC00982 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015440.1 | 1 | 269 | |||
NR_024371.1 | 1 | 269 | ||||
GenBank mRNA | BC029785.1 | - | 1 | 313 | ||
BM931404.1 | - | 1 | 271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
