siRNA Details
Gene Symbol: WDFY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014991.4 | NP_055806.2 | 48 | 8148 | ||
XM_005262858.4 | XP_005262915.1 | 50 | 8189 | |||
XM_011531757.2 | XP_011530059.1 | 49 | 8100 | |||
XM_011531759.2 | XP_011530061.1 | 48 | 8062 | |||
XM_011531760.2 | XP_011530062.1 | 48 | 8000 | |||
XM_011531761.2 | XP_011530063.1 | 49 | 8055 | |||
XM_011531762.2 | XP_011530064.1 | 48 | 8049 | |||
XM_011531763.2 | XP_011530065.1 | 47 | 7946 | |||
XM_011531764.2 | XP_011530066.1 | 48 | 8016 | |||
XM_011531765.2 | XP_011530067.1 | 45 | 7751 | |||
XM_017007906.1 | XP_016863395.1 | 51 | 8317 | |||
XM_017007907.1 | XP_016863396.1 | 51 | 8314 | |||
XM_017007908.1 | XP_016863397.1 | 50 | 8246 | |||
XM_017007909.1 | XP_016863398.1 | 36 | 5942 | |||
GenBank mRNA | AB023210.2 | - | 13 | 1831 | ||
AF538685.1 | - | 47 | 8040 | |||
AK094910.1 | - | 4 | 549 | |||
BC150246.1 | - | 13 | 1852 | |||
BX648077.1 | - | 5 | 713 | |||
BX648153.1 | - | 8 | 1041 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
