siRNA Details
Gene Symbol: WDFY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014991.4 | NP_055806.2 | 53, 54 | 8631 | ||
XM_005262858.4 | XP_005262915.1 | 55, 56 | 8672 | |||
XM_011531757.2 | XP_011530059.1 | 54, 55 | 8583 | |||
XM_011531759.2 | XP_011530061.1 | 53, 54 | 8545 | |||
XM_011531760.2 | XP_011530062.1 | 53, 54 | 8483 | |||
XM_011531761.2 | XP_011530063.1 | 54, 55 | 8538 | |||
XM_011531762.2 | XP_011530064.1 | 53, 54 | 8532 | |||
XM_011531763.2 | XP_011530065.1 | 52, 53 | 8429 | |||
XM_011531764.2 | XP_011530066.1 | 53, 54 | 8499 | |||
XM_011531765.2 | XP_011530067.1 | 50, 51 | 8234 | |||
XM_017007906.1 | XP_016863395.1 | 56, 57 | 8800 | |||
XM_017007907.1 | XP_016863396.1 | 56, 57 | 8797 | |||
XM_017007908.1 | XP_016863397.1 | 55, 56 | 8729 | |||
XM_017007909.1 | XP_016863398.1 | 41, 42 | 6425 | |||
GenBank mRNA | AB023210.2 | - | 18, 19 | 2314 | ||
AF538685.1 | - | 52, 53 | 8523 | |||
AK055806.1 | - | 4, 5 | 278 | |||
AK094910.1 | - | 9, 10 | 1032 | |||
BC150246.1 | - | 18, 19 | 2335 | |||
BX648077.1 | - | 10, 11 | 1196 | |||
BX648153.1 | - | 13, 14 | 1524 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
