siRNA Details
Gene Symbol: KLF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160124.1 | NP_001153596.1 | 2 | 437 | ||
NM_001160125.1 | NP_001153597.1 | 2 | 437 | |||
NM_001300.5 | NP_001291.3 | 2 | 437 | |||
NR_027653.1 | 2 | 437 | ||||
GenBank mRNA | AB017493.1 | - | 2 | 287 | ||
AF001461.1 | - | 2 | 200 | |||
AK293259.1 | - | 2 | 371 | |||
AK316596.1 | - | 2 | 365 | |||
BC000311.2 | - | 2 | 356 | 2821009 | ||
BC004301.1 | - | 2 | 354 | |||
BM544849.1 | - | 2 | 438 | |||
BM695496.1 | - | 1 | 13 | |||
BT006778.1 | - | 2 | 170 | |||
CR975091.1 | - | 2 | 354 | |||
CR994352.1 | - | 2 | 354 | |||
FJ619623.1 | - | 2 | 177 | |||
U44975.1 | - | 2 | 191 | |||
U51869.1 | - | 2 | 522 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
