siRNA Details
Gene Symbol: UACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008224.1 | NP_001008225.1 | 10 | 1181 | ||
NM_018003.2 | NP_060473.2 | 10 | 958 | |||
XM_005254529.4 | XP_005254586.1 | 10 | 1208 | |||
XM_011521752.2 | XP_011520054.2 | 10 | 1287 | |||
XM_011521753.2 | XP_011520055.2 | 10 | 1288 | |||
XM_011521754.2 | XP_011520056.1 | 9 | 736 | |||
XM_017022394.1 | XP_016877883.1 | 10 | 1040 | |||
XM_017022395.1 | XP_016877884.1 | 10 | 818 | |||
GenBank mRNA | AB046781.2 | - | 10 | 854 | ||
AF322916.1 | - | 10 | 903 | |||
AK055435.1 | - | 10 | 1208 | |||
AL834243.1 | - | 10 | 1183 | |||
BC016881.1 | - | 10 | 1170 | |||
BC113407.1 | - | 10 | 877 | 8322459 | ||
BC113409.1 | - | 10 | 877 | 8322461 | ||
BC143266.1 | - | 10 | 877 | 9051768 | ||
BC143267.1 | - | 7 | 550 | 9051769 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
