siRNA Details
Gene Symbol: NFKBIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005474.2 | NP_001005474.1 | 4, 5 | 443 | ||
NM_031419.3 | NP_113607.1 | 3, 4 | 576 | |||
GenBank mRNA | AB037925.1 | - | 3, 4 | 509 | ||
AF548362.1 | - | 4, 5 | 440 | |||
AK054787.1 | - | 4, 5 | 312 | |||
AK091782.1 | - | 3, 4 | 576 | |||
BC014587.2 | - | 223 | ||||
BC060800.1 | - | 3, 4 | 576 | 30344780 | ||
DA541852.1 | - | 4, 5 | 443 | |||
DQ224339.1 | - | 3, 4 | 365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
