siRNA Details
Gene Symbol: NFKBIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005474.2 | NP_001005474.1 | 5, 6 | 542 | ||
NM_031419.3 | NP_113607.1 | 4, 5 | 675 | |||
GenBank mRNA | AB037925.1 | - | 4, 5 | 608 | ||
AF548362.1 | - | 5, 6 | 539 | |||
AK054787.1 | - | 5, 6 | 411 | |||
AK091782.1 | - | 4, 5 | 675 | |||
BC014587.2 | - | 322 | ||||
BC060800.1 | - | 4, 5 | 675 | 30344780 | ||
DA541852.1 | - | 5, 6 | 542 | |||
DQ224339.1 | - | 4, 5 | 464 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
