siRNA Details
Gene Symbol: FAM87B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103536.1 | 2 | 1791 | |||
GenBank mRNA | AK097327.1 | - | 2 | 1791 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
