siRNA Details
Gene Symbol: DEFA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005217.3 | NP_005208.1 | 2 | 212 | ||
XM_011534741.2 | XP_011533043.1 | 3 | 298 | |||
GenBank mRNA | BC027917.1 | - | 2 | 199 | 5226993 | |
BC119706.2 | - | 2 | 197 | 40117146 | ||
M21131.1 | - | 1 | 41 | |||
M23281.1 | - | 2 | 177 | |||
X13621.1 | - | 2 | 216 |
Gene Symbol: DEFA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004084.3 | NP_004075.1 | 2 | 219 | ||
XM_011534740.2 | XP_011533042.1 | 3 | 290 | |||
GenBank mRNA | BC069423.1 | - | 2 | 177 | 7262694 | |
BC093791.1 | - | 2 | 174 | 7939636 | ||
BC112188.1 | - | 2 | 174 | 8327656 | ||
M21130.1 | - | 2 | 177 | |||
M26602.1 | - | 2 | 220 | |||
X52053.1 | - | 2 | 199 |
Gene Symbol: DEFA1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042500.1 | NP_001035965.1 | 2 | 219 | ||
NM_001302265.1 | NP_001289194.1 | 3 | 263 | |||
GenBank mRNA | HY073456.1 | - | 3 | 261 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
