siRNA Details
Gene Symbol: IFIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270927.1 | NP_001257856.1 | 3 | 420 | ||
NM_001270928.1 | NP_001257857.1 | 3 | 437 | |||
NM_001270929.1 | NP_001257858.1 | 3 | 432 | |||
NM_001270930.1 | NP_001257859.1 | 3 | 558 | |||
NM_001548.4 | NP_001539.3 | 2 | 323 | |||
GenBank mRNA | AK092813.1 | - | 2 | 1351 | ||
AK308688.1 | - | 3 | 324 | |||
AK314588.1 | - | 2 | 246 | |||
AL711085.1 | - | 3 | 333 | |||
BC007091.1 | - | 2 | 248 | 4250452 | ||
BI670242.1 | - | 2 | 331 | |||
BT006667.1 | - | 1 | 156 | |||
CD106320.1 | - | 2 | 199 | |||
DA822484.1 | - | 2 | 305 | |||
DB003897.1 | - | 3 | 357 | |||
DB180094.1 | - | 3 | 367 | |||
DB474377.1 | - | 3 | 467 | |||
X03557.1 | - | 2 | 220 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
