siRNA Details
Gene Symbol: SETD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007259.2 | NP_001007260.1 | 7 | 974 | ||
NM_001007261.2 | NP_001007262.1 | 8 | 1068 | |||
NM_001286752.1 | NP_001273681.1 | 8 | 1457 | |||
NM_017438.4 | NP_059134.1 | 7 | 1314 | |||
NR_040087.2 | 6 | 1225 | ||||
XM_011529636.1 | XP_011527938.1 | 7 | 1363 | |||
XM_011529637.1 | XP_011527939.1 | 7 | 1428 | |||
XM_011529638.2 | XP_011527940.1 | 7 | 1336 | |||
XM_011529639.1 | XP_011527941.1 | 7 | 1114 | |||
XM_011529640.2 | XP_011527942.1 | 7 | 1379 | |||
XM_011529642.1 | XP_011527944.1 | 8 | 1457 | |||
XM_011529643.1 | XP_011527945.1 | 8 | 1408 | |||
XM_011529644.1 | XP_011527946.1 | 8 | 969 | |||
XM_017028403.1 | XP_016883892.1 | 7 | 905 | |||
XM_017028404.1 | XP_016883893.1 | 8 | 1013 | |||
XM_017028405.1 | XP_016883894.1 | 7 | 1314 | |||
GenBank mRNA | AB004848.1 | - | 8 | 1021 | ||
AB004849.1 | - | 2 | 524 | |||
AB004852.1 | - | 6 | 845 | |||
AB004853.1 | - | 8 | 1017 | |||
AF391112.1 | - | 6 | 792 | |||
AK001660.1 | - | 7 | 881 | |||
AK095215.1 | - | 6 | 2181 | |||
AK300009.1 | - | 6 | 925 | |||
BC002898.2 | - | 7 | 913 | 3940849 | ||
BC036556.2 | - | 8 | 1131 | 5266627 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
