siRNA Details
Gene Symbol: C3orf35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302831.1 | NP_001289760.1 | 1 | 133 | ||
NM_001302832.1 | NP_001289761.1 | 1 | 133 | |||
NM_178339.2 | NP_848029.2 | 2 | 316 | |||
NM_178342.2 | NP_848032.1 | 2 | 316 | |||
NM_178344.2 | NP_848034.2 | 2 | 235 | |||
NR_126512.1 | 2 | 316 | ||||
NR_126513.1 | 2 | 316 | ||||
NR_126514.1 | 2 | 316 | ||||
NR_126515.1 | 2 | 316 | ||||
XM_017006291.1 | XP_016861780.1 | 5 | 1409 | |||
XM_017006292.1 | XP_016861781.1 | 4 | 1675 | |||
XM_017006293.1 | XP_016861782.1 | 3 | 5941 | |||
XM_017006294.1 | XP_016861783.1 | 6 | 712 | |||
XM_017006295.1 | XP_016861784.1 | 3 | 3088 | |||
XM_017006296.1 | XP_016861785.1 | 4 | 3209 | |||
GenBank mRNA | AJ493599.1 | - | 2 | 632 | ||
AJ493600.1 | - | 2 | 632 | |||
AJ493601.1 | - | 2 | 632 | |||
AJ493602.1 | - | 2 | 632 | |||
AJ493603.1 | - | 2 | 223 | |||
AJ493604.1 | - | 2 | 301 | |||
AJ493605.1 | - | 2 | 235 | |||
BC100859.2 | - | 4 | 499 | |||
BC100861.1 | - | 3 | 378 | |||
BC100862.2 | - | 4 | 499 | |||
BC112433.1 | - | 2 | 195 | |||
BC113895.1 | - | 3 | 378 | |||
BC132691.1 | - | 1 | 62 | 40146713 | ||
BC133024.1 | - | 1 | 62 | 40147046 | ||
BC144376.1 | - | 1 | 62 | 9052897 | ||
BC144377.1 | - | 1 | 62 | 9052898 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
