siRNA Details
Gene Symbol: OR5L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004739.1 | NP_001004739.1 | 1 | 113 |
Gene Symbol: OR5L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004738.1 | NP_001004738.1 | 1 | 113 | ||
GenBank mRNA | BC109249.1 | - | 1 | 113 | 40022949 | |
BC109250.1 | - | 1 | 113 | 40022959 | ||
BC136939.1 | - | 1 | 213 | 9020931 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
