siRNA Details
Gene Symbol: AKAP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008534.1 | NP_001008534.1 | 2 | 242 | ||
NM_001008535.1 | NP_001008535.1 | 2 | 242 | |||
NM_178813.5 | NP_848928.1 | 2 | 242 | |||
GenBank mRNA | AF514780.1 | - | 2 | 189 | ||
AF514781.1 | - | 2 | 189 | |||
AF514782.1 | - | 2 | 189 | |||
BC066357.1 | - | 3 | 379 | 5295545 | ||
BG208671.1 | - | 1 | 166 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
