siRNA Details
Gene Symbol: C17orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181655.3 | NP_858041.2 | 3 | 674 | ||
NM_181656.4 | NP_858042.2 | 3 | 817 | |||
GenBank mRNA | AK026583.1 | - | 3 | 755 | ||
BC021702.1 | - | 2 | 436 | |||
BC036760.1 | - | 3 | 549 | 5587712 | ||
BC105124.1 | - | 3 | 493 | 8144127 | ||
BC112073.1 | - | 3 | 505 | 8327541 | ||
BX364842.2 | - | 3 | 816 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
