siRNA Details
Gene Symbol: ZNF398 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020781.3 | NP_065832.1 | 7 | 2444 | ||
NM_170686.2 | NP_733787.1 | 6 | 2381 | |||
XM_011516439.2 | XP_011514741.1 | 7 | 2262 | |||
XM_011516440.2 | XP_011514742.1 | 6 | 2234 | |||
XM_011516441.2 | XP_011514743.1 | 6 | 2217 | |||
GenBank mRNA | AB037760.1 | - | 2 | 1402 | ||
AK290499.1 | - | 6 | 2349 | |||
AK304369.1 | - | 3 | 1536 | |||
AK304603.1 | - | 4 | 1675 | |||
AY049743.1 | - | 7 | 2444 | |||
AY049744.1 | - | 6 | 2123 | |||
BC043295.2 | - | 6 | 2369 | 5298104 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
