siRNA Details
Gene Symbol: OPTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008211.1 | NP_001008212.1 | 5 | 650 | ||
NM_001008212.1 | NP_001008213.1 | 4 | 581 | |||
NM_001008213.1 | NP_001008214.1 | 5 | 635 | |||
NM_021980.4 | NP_068815.2 | 3 | 429 | |||
GenBank mRNA | AF061034.1 | - | 5 | 523 | ||
AF420371.1 | - | 5 | 507 | |||
AF420372.1 | - | 3 | 286 | |||
AF420373.1 | - | 4 | 438 | |||
AK055403.1 | - | 4 | 870 | |||
AK303715.1 | - | 3 | 447 | |||
AK316171.1 | - | 3 | 469 | |||
BC013876.2 | - | 3 | 342 | 3831267 | ||
BC032762.1 | - | 5 | 555 | 3457195 | ||
KU178502.1 | - | 2 | 197 | |||
KU178503.1 | - | 2 | 197 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
