siRNA Details
Gene Symbol: OPTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008211.1 | NP_001008212.1 | 12 | 1671 | ||
NM_001008212.1 | NP_001008213.1 | 11 | 1602 | |||
NM_001008213.1 | NP_001008214.1 | 12 | 1656 | |||
NM_021980.4 | NP_068815.2 | 10 | 1450 | |||
GenBank mRNA | AF061034.1 | - | 12 | 1544 | ||
AF420371.1 | - | 12 | 1528 | |||
AF420372.1 | - | 10 | 1307 | |||
AF420373.1 | - | 11 | 1459 | |||
AK055403.1 | - | 11 | 1891 | |||
AK122746.1 | - | 2 | 1343 | |||
AK303715.1 | - | 10 | 1468 | |||
AK316171.1 | - | 10 | 1490 | |||
BC013876.2 | - | 10 | 1363 | 3831267 | ||
BC032762.1 | - | 12 | 1558 | 3457195 | ||
KU178502.1 | - | 9 | 1218 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
