siRNA Details
Gene Symbol: TCEAL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006684.1 | NP_001006685.1 | 2 | 419 | ||
NM_153333.2 | NP_699164.1 | 3 | 492 | |||
GenBank mRNA | AK026349.1 | - | 3 | 469 | ||
AL833632.1 | - | 2 | 412 | |||
BC035573.1 | - | 3 | 509 | 5405281 | ||
BU932815.1 | - | 3 | 503 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001168578, NM_025703, AK156414, AK012030, AK027927, and others.
Gene Ontology Categories:
