siRNA Details
Gene Symbol: TCEAL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006684.1 | NP_001006685.1 | 2 | 343 | ||
NM_153333.2 | NP_699164.1 | 3 | 416 | |||
GenBank mRNA | AK026349.1 | - | 3 | 393 | ||
AL833632.1 | - | 2 | 336 | |||
BC035573.1 | - | 3 | 433 | 5405281 | ||
BU932815.1 | - | 3 | 428 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
