siRNA Details
Gene Symbol: ISCU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301140.1 | NP_001288069.1 | 6 | 1863 | ||
NM_001301141.1 | NP_001288070.1 | 6 | 1874 | |||
NM_001320042.1 | NP_001306971.1 | 5 | 2950 | |||
NM_014301.4 | NP_055116.1 | 6 | 1873 | |||
NM_213595.3 | NP_998760.1 | 5 | 1777 | |||
NR_135127.1 | 7 | 1973 | ||||
XM_011538100.2 | XP_011536402.1 | 6 | 3046 | |||
XM_011538101.2 | XP_011536403.1 | 4 | 1698 | |||
GenBank mRNA | AK000574.1 | - | 5 | 620 | ||
AK022758.1 | - | 4 | 3374 | |||
AK057251.1 | - | 6 | 1874 | |||
AY009127.1 | - | 6 | 728 | |||
AY009128.1 | - | 5 | 632 | |||
BC011906.2 | - | 5 | 633 | 4136262 | ||
BC061903.1 | - | 5 | 626 | 4183674 | ||
BG621612.1 | - | 244 | ||||
BG702203.1 | - | 6 | 714 | |||
BM423850.1 | - | 5 | 623 | |||
BM921073.1 | - | 5 | 718 | |||
BU602914.1 | - | 5 | 639 | |||
EU329002.1 | - | 7 | 830 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
