siRNA Details
Gene Symbol: UBC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021009.6 | NP_066289.3 | 2 | 702 | ||
GenBank mRNA | AB009010.1 | - | 2 | 312 | ||
AB209782.1 | - | 292 | ||||
AK026846.1 | - | 2 | 323 | |||
AK056939.1 | - | 3 | 677 | |||
AK097415.1 | - | 1 | 668 | |||
AK098827.1 | - | 2 | 311 | |||
AK129749.1 | - | 2 | 311 | |||
AK291539.1 | - | 2 | 311 | |||
AY819759.1 | - | 1 | 262 | |||
BC000449.2 | - | 2 | 297 | |||
BC008955.2 | - | 2 | 285 | |||
BC039193.1 | - | 2 | 312 | 4076286 | ||
BC080583.1 | - | 2 | 285 | |||
BC093445.1 | - | 2 | 311 | |||
BU195276.1 | - | 2 | 297 | |||
M26880.1 | - | 379 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
