siRNA Details
Gene Symbol: RBPMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008710.2 | NP_001008710.1 | 5 | 880 | ||
NM_001008711.2 | NP_001008711.1 | 5 | 880 | |||
NM_001008712.2 | NP_001008712.1 | 5 | 880 | |||
NM_006867.3 | NP_006858.1 | 5 | 880 | |||
XM_017012980.1 | XP_016868469.1 | 5 | 875 | |||
XM_017012981.1 | XP_016868470.1 | 5 | 875 | |||
XM_017012982.1 | XP_016868471.1 | 5 | 875 | |||
XM_017012983.1 | XP_016868472.1 | 5 | 1861 | |||
XM_017012984.1 | XP_016868473.1 | 5 | 875 | |||
XM_017012985.1 | XP_016868474.1 | 2 | 304 | |||
XM_017012986.1 | XP_016868475.1 | 5 | 875 | |||
XM_017012987.1 | XP_016868476.1 | 5 | 875 | |||
XM_017012988.1 | XP_016868477.1 | 5 | 875 | |||
XM_017012989.1 | XP_016868478.1 | 5 | 875 | |||
XM_017012990.1 | XP_016868479.1 | 5 | 875 | |||
XM_017012991.1 | XP_016868480.1 | 6 | 1022 | |||
XM_017012992.1 | XP_016868481.1 | 5 | 875 | |||
GenBank mRNA | AK304858.1 | - | 6 | 698 | ||
AK310533.1 | - | 5 | 308 | |||
BC003608.2 | - | 5 | 821 | 3505124 | ||
BC063494.1 | - | 5 | 287 | |||
BC092476.1 | - | 5 | 368 | 4837343 | ||
D84107.1 | - | 5 | 854 | |||
D84108.1 | - | 5 | 854 | |||
D84109.1 | - | 5 | 854 | |||
D84110.1 | - | 5 | 854 | |||
D84111.1 | - | 3 | 154 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
