siRNA Details
Gene Symbol: SERHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027786.1 | 9 | 730 | |||
GenBank mRNA | AK094872.1 | - | 8 | 1086 | ||
AL365513.1 | - | 9 | 719 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
