siRNA Details
Gene Symbol: IVNS1ABP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006469.4 | NP_006460.2 | 4 | 854 | ||
XM_005244843.3 | XP_005244900.1 | 4 | 810 | |||
XM_011509079.2 | XP_011507381.1 | 4 | 864 | |||
XM_017000084.1 | XP_016855573.1 | 4 | 810 | |||
GenBank mRNA | AB020657.1 | - | 4 | 861 | ||
AF161553.1 | - | 4 | 770 | |||
AF205218.1 | - | 4 | 786 | |||
AJ012449.1 | - | 4 | 768 | |||
AK001625.1 | - | 2 | 192 | |||
AK023020.1 | - | 3 | 462 | |||
AK023123.1 | - | 4 | 798 | |||
AK292431.1 | - | 4 | 604 | |||
BC067739.1 | - | 4 | 754 | 5299055 | ||
DQ443528.1 | - | 4 | 743 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
