siRNA Details
Gene Symbol: IVNS1ABP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006469.4 | NP_006460.2 | 15 | 2471 | ||
XM_005244843.3 | XP_005244900.1 | 16 | 2568 | |||
XM_011509079.2 | XP_011507381.1 | 15 | 2502 | |||
XM_011509080.2 | XP_011507382.1 | 14 | 2336 | |||
XM_017000084.1 | XP_016855573.1 | 16 | 2589 | |||
GenBank mRNA | AB020657.1 | - | 15 | 2478 | ||
AF161553.1 | - | 15 | 2384 | |||
AF205218.1 | - | 15 | 2403 | |||
AJ012449.1 | - | 15 | 2384 | |||
AK000931.1 | - | 8 | 1111 | |||
AK001273.1 | - | 9 | 1302 | |||
AK001625.1 | - | 13 | 1809 | |||
AK001824.1 | - | 8 | 1189 | |||
AK023020.1 | - | 15 | 2220 | |||
AK023123.1 | - | 16 | 2552 | |||
AK092912.1 | - | 6 | 1471 | |||
AK093912.1 | - | 8 | 1988 | |||
AK292431.1 | - | 15 | 2221 | |||
BC067739.1 | - | 15 | 2371 | 5299055 | ||
BX648273.1 | - | 8 | 2048 | |||
DQ443528.1 | - | 15 | 2360 | |||
DQ443529.1 | - | 11 | 1515 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
