siRNA Details
Gene Symbol: IVNS1ABP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006469.4 | NP_006460.2 | 15 | 3354 | ||
XM_005244843.3 | XP_005244900.1 | 16 | 3451 | |||
XM_011509079.2 | XP_011507381.1 | 15 | 3385 | |||
XM_011509080.2 | XP_011507382.1 | 14 | 3219 | |||
XM_017000084.1 | XP_016855573.1 | 16 | 3472 | |||
GenBank mRNA | AB020657.1 | - | 15 | 3361 | ||
AF205218.1 | - | 15 | 3286 | |||
AK000931.1 | - | 8 | 1995 | |||
AK001625.1 | - | 13 | 2692 | |||
AK001824.1 | - | 8 | 2072 | |||
AK023123.1 | - | 16 | 3435 | |||
AL137640.1 | - | 1 | 682 | |||
BX648273.1 | - | 8 | 2929 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
