siRNA Details
Gene Symbol: GP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007240.2 | NP_001007241.2 | 8 | 1241 | ||
NM_001007241.2 | NP_001007242.2 | 7 | 800 | |||
NM_001007242.2 | NP_001007243.2 | 6 | 791 | |||
NM_001502.3 | NP_001493.2 | 7 | 1232 | |||
XM_017023155.1 | XP_016878644.1 | 7 | 985 | |||
GenBank mRNA | AB035541.1 | - | 6 | 1021 | ||
AB035542.1 | - | 5 | 580 | |||
AK293384.1 | - | 7 | 988 | |||
AK312277.1 | - | 7 | 1119 | |||
BC032693.2 | - | 5 | 638 | 5229838 | ||
BX647831.1 | - | 8 | 1129 | |||
BX648023.1 | - | 7 | 1126 | |||
BX648201.1 | - | 6 | 678 | |||
BX649067.1 | - | 7 | 687 | |||
CR749600.1 | - | 7 | 1191 | |||
D38225.1 | - | 6 | 592 | |||
U36221.1 | - | 7 | 1101 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
