siRNA Details
Gene Symbol: GP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007240.2 | NP_001007241.2 | 8, 9 | 1458 | ||
NM_001007241.2 | NP_001007242.2 | 7, 8 | 1017 | |||
NM_001007242.2 | NP_001007243.2 | 6, 7 | 1008 | |||
NM_001502.3 | NP_001493.2 | 7, 8 | 1449 | |||
XM_017023155.1 | XP_016878644.1 | 7, 8 | 1202 | |||
GenBank mRNA | AK293384.1 | - | 7, 8 | 1205 | ||
AK312277.1 | - | 7, 8 | 1336 | |||
BC032693.2 | - | 5, 6 | 855 | 5229838 | ||
D38225.1 | - | 6, 7 | 809 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
