siRNA Details
Gene Symbol: WDR31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006615.2 | NP_001006616.1 | 8, 9 | 959 | ||
NM_001012361.3 | NP_001012361.1 | 9, 10 | 1092 | |||
NM_145241.4 | NP_660284.1 | 9, 10 | 1089 | |||
NR_134677.1 | 9, 10 | 1060 | ||||
XM_011518194.2 | XP_011516496.1 | 8, 9 | 1003 | |||
GenBank mRNA | AK074891.1 | - | 8, 9 | 873 | ||
AK093240.1 | - | 9, 10 | 1021 | |||
AK313843.1 | - | 9, 10 | 1031 | |||
AL833635.1 | - | 9, 10 | 977 | |||
BC012352.1 | - | 9, 10 | 1021 | 4634445 | ||
BC022982.1 | - | 9, 10 | 1018 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_023597, NM_001290522, NM_001290521, AF274321, BC026500, and others.
