siRNA Details
Gene Symbol: METTL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007239.1 | NP_001007240.1 | 8 | 1939 | ||
NM_014955.2 | NP_055770.1 | 8 | 2082 | |||
NM_015935.4 | NP_057019.3 | 8 | 2407 | |||
XM_017001457.1 | XP_016856946.1 | 6 | 1682 | |||
XM_017001458.1 | XP_016856947.1 | 7 | 2359 | |||
XM_017001459.1 | XP_016856948.1 | 7 | 1909 | |||
GenBank mRNA | AB020666.2 | - | 8 | 2162 | ||
AB242174.1 | - | 8 | 2332 | |||
AB242175.1 | - | 8 | 2320 | |||
AK001172.1 | - | 6 | 1094 | |||
AK027552.1 | - | 8 | 2326 | |||
AK027621.1 | - | 8 | 2021 | |||
AK027886.1 | - | 7 | 1878 | |||
AK074552.1 | - | 4 | 1160 | |||
AK094818.1 | - | 1 | 1447 | |||
AK124482.1 | - | 1 | 2128 | |||
AK129561.1 | - | 7 | 1755 | |||
AK223493.1 | - | 8 | 2368 | |||
AK291740.1 | - | 8 | 2354 | |||
AK304471.1 | - | 9 | 2188 | |||
AL049669.1 | - | 8 | 1864 | |||
BC006814.1 | - | 5 | 957 | |||
BC029083.1 | - | 8 | 2161 | 4509086 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
