siRNA Details
Gene Symbol: METTL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007239.1 | NP_001007240.1 | 8 | 2265 | ||
NM_014955.2 | NP_055770.1 | 8 | 2408 | |||
NM_015935.4 | NP_057019.3 | 8 | 2733 | |||
XM_017001457.1 | XP_016856946.1 | 6 | 2008 | |||
XM_017001458.1 | XP_016856947.1 | 7 | 2685 | |||
XM_017001459.1 | XP_016856948.1 | 7 | 2235 | |||
GenBank mRNA | AB242175.1 | - | 8 | 2646 | ||
AK027621.1 | - | 8 | 2347 | |||
AK094818.1 | - | 1 | 1773 | |||
AK129561.1 | - | 7 | 2081 | |||
AK291740.1 | - | 8 | 2680 | |||
AL049669.1 | - | 8 | 2190 | |||
BX094025.1 | - | 1 | 55 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
