siRNA Details
Gene Symbol: KIR2DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014219.2 | NP_055034.2 | 4 | 532 | ||
GenBank mRNA | AF285434.1 | - | 3 | 463 | ||
AY366242.1 | - | 4 | 532 | |||
KP260924.1 | - | 4 | 499 | |||
L76669.1 | - | 4 | 514 | |||
U24075.1 | - | 4 | 517 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
