siRNA Details
Gene Symbol: KIR2DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014219.2 | NP_055034.2 | 8 | 1229 | ||
GenBank mRNA | AY366242.1 | - | 8 | 1229 | ||
L76669.1 | - | 8 | 1211 | |||
U24075.1 | - | 8 | 1214 |
Gene Symbol: KIR2DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF285431.1 | - | 8 | 1233 | ||
AY366237.1 | - | 8 | 1236 | |||
AY366238.1 | - | 8 | 1182 | |||
AY366239.1 | - | 8 | 1230 | |||
AY366240.1 | - | 8 | 1238 | |||
AY366241.1 | - | 8 | 1237 | |||
AY388471.1 | - | 8 | 1230 | |||
L41267.1 | - | 9 | 1232 | |||
U24076.1 | - | 9 | 1233 | |||
U24078.1 | - | 8 | 1240 |
Gene Symbol: KIR3DS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083539.2 | NP_001077008.1 | 10 | 1524 | ||
NM_001282170.1 | NP_001269099.1 | 9 | 1239 | |||
NM_001282171.1 | NP_001269100.1 | 8 | 1203 | |||
GenBank mRNA | AK301140.1 | - | 10 | 1524 | ||
L76661.1 | - | 10 | 1497 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
