siRNA Details
Gene Symbol: KIR2DL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AY366239.1 | - | 8 | 1183 | ||
AY366240.1 | - | 8 | 1191 | |||
AY366241.1 | - | 8 | 1190 | |||
AY388471.1 | - | 8 | 1183 | |||
L41267.1 | - | 9 | 1185 | |||
U24076.1 | - | 9 | 1186 |
Gene Symbol: KIR2DL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014219.2 | NP_055034.2 | 8 | 1182 | ||
GenBank mRNA | AY366242.1 | - | 8 | 1182 | ||
L76669.1 | - | 8 | 1164 | |||
U24075.1 | - | 8 | 1167 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
