siRNA Details
Gene Symbol: FOXD4L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199135.4 | NP_954586.4 | 1 | 2102 | ||
GenBank mRNA | AY344642.1 | - | 1 | 2102 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
