siRNA Details
Gene Symbol: SLC25A25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006641.3 | NP_001006642.1 | 10 | 3351 | ||
NM_001006642.3 | NP_001006643.1 | 10 | 3596 | |||
NM_001265614.2 | NP_001252543.1 | 11 | 3632 | |||
NM_052901.4 | NP_443133.2 | 10 | 3214 | |||
NR_049766.2 | 10 | 3257 | ||||
XM_005251688.1 | XP_005251745.1 | 11 | 3387 | |||
XM_005251689.4 | XP_005251746.1 | 10 | 3610 | |||
XM_006716948.2 | XP_006717011.1 | 11 | 3788 | |||
XM_006716949.3 | XP_006717012.1 | 11 | 3648 | |||
GenBank mRNA | AB067483.1 | - | 11 | 3631 | ||
AJ619963.1 | - | 10 | 3515 | |||
AJ619989.1 | - | 10 | 3139 | |||
AJ619990.1 | - | 10 | 3234 | |||
AJ619991.1 | - | 10 | 3196 | |||
AJ619992.1 | - | 10 | 3182 | |||
AY358515.1 | - | 10 | 3189 | |||
BC005163.2 | - | 6 | 2648 | |||
BC044631.1 | - | 1 | 1774 | |||
BC061925.1 | - | 1 | 771 | |||
BC089448.1 | - | 10 | 3205 | 30558551 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
