siRNA Details
Gene Symbol: EXPH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308019.1 | NP_001294948.1 | 2, 3 | 100 | ||
NM_015065.2 | NP_055880.2 | 1, 2 | 227 | |||
XM_017017397.1 | XP_016872886.1 | 2, 3 | 116 | |||
XM_017017398.1 | XP_016872887.1 | 2, 3 | 116 | |||
GenBank mRNA | AB014524.1 | - | 2, 3 | 100 | ||
AY099469.1 | - | 1, 2 | 213 | |||
BC113119.1 | - | 1, 2 | 198 | 40037748 | ||
BC117701.1 | - | 1, 2 | 198 | 40074153 | ||
BP358360.1 | - | 1, 2 | 227 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_176846, BC120905, AB098163.
Gene Ontology Categories:
