siRNA Details
Gene Symbol: TSPAN17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006616.2 | NP_001006617.2 | 4 | 614 | ||
NM_012171.2 | NP_036303.1 | 4 | 614 | |||
NM_130465.4 | NP_569732.2 | 4 | 614 | |||
XM_005265876.3 | XP_005265933.1 | 2 | 396 | |||
GenBank mRNA | AK024427.1 | - | 4 | 504 | ||
AY037146.1 | - | 2 | 336 | |||
BC010405.2 | - | 4 | 436 | 3621871 | ||
BC067105.1 | - | 4 | 499 | 6646576 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
