siRNA Details
Gene Symbol: SCRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031309.5 | NP_112599.2 | 2 | 3509 | ||
GenBank mRNA | AL512683.1 | - | 1 | 1024 | ||
BC024967.2 | - | 1 | 1030 |

More Information
Other Species Targeted

Gene Ontology Categories:
