siRNA Details
Gene Symbol: TULP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007466.2 | NP_001007467.1 | 13 | 4667 | ||
NM_020245.4 | NP_064630.2 | 14 | 7168 | |||
XM_011535946.1 | XP_011534248.1 | 16 | 7914 | |||
XM_017011063.1 | XP_016866552.1 | 17 | 8063 | |||
XM_017011064.1 | XP_016866553.1 | 18 | 8228 | |||
XM_017011065.1 | XP_016866554.1 | 17 | 7981 | |||
XM_017011066.1 | XP_016866555.1 | 16 | 7871 | |||
XM_017011067.1 | XP_016866556.1 | 16 | 7964 | |||
XM_017011068.1 | XP_016866557.1 | 16 | 7930 | |||
XM_017011069.1 | XP_016866558.1 | 16 | 7997 | |||
XM_017011070.1 | XP_016866559.1 | 15 | 7845 | |||
XM_017011071.1 | XP_016866560.1 | 15 | 5496 | |||
GenBank mRNA | AB037818.1 | - | 13 | 4667 | ||
AF219946.2 | - | 14 | 7170 | |||
AF288480.1 | - | 13 | 4666 | |||
BC152476.1 | - | 13 | 4667 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
