siRNA Details
Gene Symbol: SFMBT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016329.3 | NP_057413.2 | 15, 16 | 2014 | ||
XM_005265221.3 | XP_005265278.1 | 15, 16 | 2130 | |||
XM_006713203.3 | XP_006713266.1 | 16, 17 | 2252 | |||
XM_006713204.3 | XP_006713267.1 | 15, 16 | 1952 | |||
XM_011533824.2 | XP_011532126.1 | 16, 17 | 2128 | |||
XM_011533825.2 | XP_011532127.1 | 16, 17 | 1975 | |||
GenBank mRNA | AB189472.1 | - | 14, 15 | 1638 | ||
AF168132.1 | - | 15, 16 | 1951 | |||
AK313965.1 | - | 15, 16 | 2009 | |||
AL080140.1 | - | 12, 13 | 1292 | |||
BC014614.2 | - | 15, 16 | 1898 | 3029598 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
