siRNA Details
Gene Symbol: NUMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005743.1 | NP_001005743.1 | 7 | 617 | ||
NM_001005744.1 | NP_001005744.1 | 7 | 617 | |||
NM_001005745.1 | NP_001005745.1 | 6 | 584 | |||
NM_001320114.1 | NP_001307043.1 | 7 | 637 | |||
NM_003744.5 | NP_003735.3 | 6 | 584 | |||
GenBank mRNA | AF015040.1 | - | 7 | 575 | ||
AF108092.1 | - | 5 | 402 | |||
AF171938.1 | - | 7 | 567 | |||
AF171939.1 | - | 7 | 567 | |||
AF171940.1 | - | 6 | 534 | |||
AF171941.1 | - | 6 | 534 | |||
AK023670.1 | - | 1 | 654 | |||
AK226069.1 | - | 1 | 2153 | |||
AK304193.1 | - | 5 | 420 | |||
AK315130.1 | - | 5 | 465 | |||
BC020788.1 | - | 5 | 465 | |||
BC068476.1 | - | 6 | 584 | 4822300 | ||
BX248073.1 | - | 6 | 474 | |||
BX248775.1 | - | 5 | 438 | |||
BX248781.1 | - | 5 | 479 | |||
BX647137.1 | - | 7 | 576 | |||
EU265734.1 | - | 5 | 336 | |||
EU265735.1 | - | 4 | 303 | |||
EU265736.1 | - | 5 | 336 | |||
EU265737.1 | - | 4 | 303 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
