siRNA Details
Gene Symbol: ZDHHC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024630.2 | NP_078906.2 | 9 | 2688 | ||
NM_153746.1 | NP_714968.1 | 9 | 2643 | |||
XM_005267146.3 | XP_005267203.1 | 8 | 2059 | |||
XM_011536120.2 | XP_011534422.1 | 9 | 3269 | |||
XM_011536123.2 | XP_011534425.1 | 9 | 2080 | |||
XM_017011307.1 | XP_016866796.1 | 9 | 2373 | |||
XM_017011309.1 | XP_016866798.1 | 9 | 2328 | |||
XM_017011311.1 | XP_016866800.1 | 8 | 2235 | |||
XM_017011312.1 | XP_016866801.1 | 9 | 2388 | |||
XM_017011313.1 | XP_016866802.1 | 10 | 5085 | |||
GenBank mRNA | AF542388.1 | - | 9 | 2688 | ||
AF542389.1 | - | 9 | 2643 | |||
AJ420552.1 | - | 1 | 920 | |||
AK024637.1 | - | 5 | 1497 | |||
AK124573.1 | - | 6 | 5055 | |||
BC029881.1 | - | 8 | 1802 | |||
BC056399.1 | - | 3 | 1331 |
Gene Symbol: LOC100128551 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125148.1 | - | 2 | 2885 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
