siRNA Details
Gene Symbol: WDHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008396.2 | NP_001008397.1 | 5, 6 | 506 | ||
NM_007086.3 | NP_009017.1 | 6, 7 | 599 | |||
XM_006720012.2 | XP_006720075.1 | 6, 7 | 597 | |||
XM_011536373.2 | XP_011534675.1 | 5, 6 | 510 | |||
XM_017020940.1 | XP_016876429.1 | 5, 6 | 510 | |||
XM_017020941.1 | XP_016876430.1 | 3, 4 | 320 | |||
GenBank mRNA | AJ006266.1 | - | 5, 6 | 548 | ||
AK293005.1 | - | 6, 7 | 566 | |||
BC000622.2 | - | 6, 7 | 564 | |||
BC005040.1 | - | 6, 7 | 564 | |||
BC043349.1 | - | 6, 7 | 587 | |||
BC063041.1 | - | 6, 7 | 575 | 6514641 | ||
DA189308.1 | - | 5, 6 | 506 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
