siRNA Details
Gene Symbol: FAM45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303111.1 | NP_001290040.1 | 10 | 2254 | ||
NM_001303112.1 | NP_001290041.1 | 9 | 2001 | |||
NM_001303113.1 | NP_001290042.1 | 8 | 1920 | |||
NM_207009.3 | NP_996892.1 | 9 | 2049 | |||
NR_130122.1 | 8 | 1937 | ||||
XM_006717849.3 | XP_006717912.1 | 9 | 2069 | |||
XM_006717850.3 | XP_006717913.1 | 10 | 2311 | |||
XM_006717851.3 | XP_006717914.1 | 10 | 2291 | |||
XM_006717852.3 | XP_006717915.1 | 10 | 2275 | |||
XM_017016261.1 | XP_016871750.1 | 10 | 2784 | |||
XM_017016262.1 | XP_016871751.1 | 11 | 2499 | |||
XM_017016263.1 | XP_016871752.1 | 11 | 2480 | |||
XM_017016264.1 | XP_016871753.1 | 9 | 1444 | |||
XM_017016265.1 | XP_016871754.1 | 10 | 2295 | |||
XM_017016266.1 | XP_016871755.1 | 11 | 2476 | |||
XM_017016267.1 | XP_016871756.1 | 11 | 2460 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
