siRNA Details
Gene Symbol: RPL39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000.3 | NP_000991.1 | 3 | 328 | ||
GenBank mRNA | AK098280.1 | - | 2 | 1826 | ||
BC001019.2 | - | 1 | 280 | 3343512 | ||
BC070205.1 | - | 1 | 298 | 6732169 | ||
BX476041.1 | - | 3 | 329 | |||
D79205.1 | - | 3 | 282 | |||
U57846.1 | - | 3 | 263 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
