siRNA Details
Gene Symbol: SPANXB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032461.3 | NP_115850.2 | 2 | 392 | ||
GenBank mRNA | AF098307.1 | - | 2 | 392 | ||
AJ457796.1 | - | 2 | 392 | |||
BC034472.1 | - | 2 | 390 | 4824918 | ||
BC137533.1 | - | 2 | 382 | 9021533 | ||
BC137536.1 | - | 2 | 382 | 9021536 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
