siRNA Details
Gene Symbol: KIAA0100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014680.3 | NP_055495.2 | 11 | 1310 | ||
XM_005258073.2 | XP_005258130.1 | 11 | 1305 | |||
XM_017025423.1 | XP_016880912.1 | 10 | 1159 | |||
GenBank mRNA | AK130007.1 | - | 11 | 1265 | ||
BC048096.1 | - | 9 | 1818 | |||
BC050440.1 | - | 11 | 1306 | |||
BC120873.1 | - | 11 | 1169 | 40075102 | ||
CR933634.1 | - | 11 | 1262 | |||
D43947.3 | - | 11 | 1282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
